(2',4'-Difluoro-[1,1'-biphenyl]-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

61056-500MG 61056-1G 61056-5G
77336-110EA 375.95 USD
77336-110 77355-336 77379-396
(2',4'-Difluoro-[1,1'-biphenyl]-3-yl)methanol ≥97%
(2',4'-Difluoro-[1,1'-biphenyl]-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
MW: 220.2147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31649846
CAS Number: 1477478-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR