3-Chloro-4-methoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61049-500MG 61049-1G 61049-5G 61049-25G
77331-104EA 275.01 USD
77331-104 77355-332 77367-300 77388-824
3-Chloro-4-methoxy-2-methylbenzoic acid ≥97%
3-Chloro-4-methoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.6189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760115
CAS Number: 2385636-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR