3-Chloro-4-methoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61047-500MG 61047-1G 61047-5G 61047-25G
77331-102EA 226.6 USD
77331-102 77355-330 77367-298 77388-822
3-Chloro-4-methoxy-2-methylbenzaldehyde ≥97%
3-Chloro-4-methoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.6195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760113
CAS Number: 1352740-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR