2-(Benzyloxy)-3-fluoro-1,4-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

61045-500MG 61045-1G 61045-25G 61045-5G
77331-100EA 301.79 USD
77331-100 77355-328 77388-820 77379-394
2-(Benzyloxy)-3-fluoro-1,4-diiodobenzene ≥97%
2-(Benzyloxy)-3-fluoro-1,4-diiodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉FI₂O
MW: 454.0173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760111
CAS Number: 2764728-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR