2-Ethoxy-3-fluoro-1,4-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

61044-500MG 61044-1G 61044-5G 61044-25G
77331-098EA 271.92 USD
77331-098 77355-326 77367-296 77388-818
2-Ethoxy-3-fluoro-1,4-diiodobenzene ≥97%
2-Ethoxy-3-fluoro-1,4-diiodobenzene

Precaución: for Research Use Only

Formula: C₈H₇FI₂O
MW: 391.9479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28791148
CAS Number: 1806373-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR