5-Isobutoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61037-500MG 61037-1G 61037-5G 61037-25G
77331-094EA 122.57 USD
77331-094 77343-952 77367-292 77379-392
5-Isobutoxy-2-methylbenzaldehyde ≥97%
5-Isobutoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34760110
CAS Number: 2764732-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR