1,2,5-Trichloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61033-500MG 61033-1G 61033-5G 61033-25G
77331-092EA 272.95 USD
77331-092 77355-322 77367-290 77388-814
1,2,5-Trichloro-3-methylbenzene ≥97%
1,2,5-Trichloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃
MW: 195.4736 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18400165
CAS Number: 56961-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR