2,5-Dichloro-1-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61032-500MG 61032-1G 61032-5G
77338-898EA 528.39 USD
77338-898 77355-320 77379-390
2,5-Dichloro-1-iodo-3-methylbenzene ≥97%
2,5-Dichloro-1-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂I
MW: 286.9250 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786702
CAS Number: 1806283-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR