3-Chloro-4-iodo-2,6-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

61030-500MG 61030-1G 61030-5G 61030-25G
77331-088EA 176.13 USD
77331-088 77346-538 77367-286 77388-810
3-Chloro-4-iodo-2,6-dimethoxypyridine ≥97%
3-Chloro-4-iodo-2,6-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇ClINO₂
Storage Temperature: Ambient
MDL Number: MFCD34760108
CAS Number: 2764728-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR