2-Chloro-5-isopropoxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

61013-500MG 61013-1G 61013-5G 61013-25G
77331-086EA 207.03 USD
77331-086 77355-316 77367-284 77388-808
2-Chloro-5-isopropoxy-4-methylpyridine ≥97%
2-Chloro-5-isopropoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MW: 185.6506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23113487
CAS Number: 1394967-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR