5-(Benzyloxy)-2-chloro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

61012-500MG 61012-1G 61012-5G 61012-25G
77331-084EA 204.97 USD
77331-084 77355-314 77367-282 77388-806
5-(Benzyloxy)-2-chloro-4-methylpyridine ≥97%
5-(Benzyloxy)-2-chloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO
MW: 233.6934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32781969
CAS Number: 2755724-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR