5-(2,2-Dichloro-1-hydroxyethyl)-2-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

60983-1G 60983-5G 60983-25G
77341-946EA 108.15 USD
77341-946 77367-278 77379-384
5-(2,2-Dichloro-1-hydroxyethyl)-2-methoxyphenol ≥95%
5-(2,2-Dichloro-1-hydroxyethyl)-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O₃
MW: 237.0799 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760084
CAS Number: 2764731-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR