5-(6-Chloro-2,3-difluorophenyl)thiazol-2-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
60967-500MG
60967-1G
60967-5G
77336-512EA
388.31
USD
77336-512
77355-310
77379-382
5-(6-Chloro-2,3-difluorophenyl)thiazol-2-amine ≥97%
5-(6-Chloro-2,3-difluorophenyl)thiazol-2-amine
Precaución: for Research Use Only
Formula:
C₉H₅ClF₂N₂S MW: 246.6642 g/mol Storage Temperature: Ambient |
CAS Number:
1856762-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...