(2-Chloropyridin-4-yl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

60963-1G 60963-5G 60963-25G
77341-046EA 66.95 USD
77341-046 77367-272 77379-376
(2-Chloropyridin-4-yl)(thiomorpholino)methanone ≥97%
(2-Chloropyridin-4-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂OS
MW: 242.7251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11172208
CAS Number: 1042781-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR