(2-Chloropyridin-4-yl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

60961-1G 60961-5G 60961-25G
77340-770EA 60.77 USD
77340-770 77367-270 77379-374
(2-Chloropyridin-4-yl)(piperidin-1-yl)methanone ≥97%
(2-Chloropyridin-4-yl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O
MW: 224.6867 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864519
CAS Number: 90287-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR