3-Bromo-5-(tert-butyl)-2-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60925-500MG 60925-1G 60925-5G 60925-25G
77331-076EA 162.74 USD
77331-076 77346-076 77367-262 77388-798
3-Bromo-5-(tert-butyl)-2-ethoxybenzaldehyde ≥97%
3-Bromo-5-(tert-butyl)-2-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrO₂
MW: 285.1768 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760037
CAS Number: 2215467-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR