3-Bromo-5-(tert-butyl)-2-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

60909-500MG 60909-1G 60909-5G 60909-25G
77331-072EA 168.92 USD
77331-072 77346-300 77367-258 77388-796
3-Bromo-5-(tert-butyl)-2-isopropoxybenzaldehyde ≥95%
3-Bromo-5-(tert-butyl)-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₉BrO₂
MW: 299.2034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760023
CAS Number: 2217002-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR