3-Bromo-5-(tert-butyl)-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60907-500MG 60907-1G 60907-5G 60907-25G
77331-070EA 170.98 USD
77331-070 77346-380 77367-256 77388-794
3-Bromo-5-(tert-butyl)-2-isopropoxybenzoic acid ≥97%
3-Bromo-5-(tert-butyl)-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉BrO₃
MW: 315.2028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760021
CAS Number: 2216726-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR