2-Fluoro-4,5-dimethoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

60897-500MG 60897-1G 60897-5G 60897-25G
77331-066EA 187.46 USD
77331-066 77355-302 77367-250 77388-790
2-Fluoro-4,5-dimethoxy-4'-methyl-1,1'-biphenyl ≥97%
2-Fluoro-4,5-dimethoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD34760017
CAS Number: 1246661-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR