2-Fluoro-4,5-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

60896-500MG 60896-1G 60896-5G 60896-25G
77331-064EA 187.46 USD
77331-064 77355-300 77367-248 77388-788
2-Fluoro-4,5-dimethoxy-1,1'-biphenyl ≥97%
2-Fluoro-4,5-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO₂
MW: 232.2502 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760016
CAS Number: 2755719-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR