3-Bromo-5-(tert-butyl)-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60879-500MG 60879-1G 60879-5G 60879-25G
77331-062EA 184.37 USD
77331-062 77347-244 77367-246 77388-786
3-Bromo-5-(tert-butyl)-2-hydroxybenzoic acid ≥97%
3-Bromo-5-(tert-butyl)-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₃
MW: 273.1231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17167528
CAS Number: 73469-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR