Methyl 5-(tert-butyl)-2-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60864-500MG 60864-1G 60864-5G
77338-876EA 527.36 USD
77338-876 77355-296 77379-364
Methyl 5-(tert-butyl)-2-ethoxybenzoate ≥97%
Methyl 5-(tert-butyl)-2-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
MW: 236.3068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31771845
CAS Number: 1369363-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR