Methyl 5-(tert-butyl)-2-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60862-500MG 60862-1G 60862-5G
77335-868EA 367.71 USD
77335-868 77355-294 77379-362
Methyl 5-(tert-butyl)-2-isopropoxybenzoate ≥97%
Methyl 5-(tert-butyl)-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₂O₃
MW: 250.3333 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31771667
CAS Number: 1368216-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR