2-Ethoxy-3-fluoro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

60852-500MG 60852-1G 60852-5G
77340-118EA 766.32 USD
77340-118 77355-288 77379-358
2-Ethoxy-3-fluoro-6-methoxybenzoic acid ≥97%
2-Ethoxy-3-fluoro-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
Storage Temperature: Ambient
MDL Number: MFCD34759989
CAS Number: 2386035-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR