2-Ethoxy-1-fluoro-3-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

60849-500MG 60849-1G 60849-25G 60849-5G
77331-056EA 305.91 USD
77331-056 77355-284 77388-780 77379-354
2-Ethoxy-1-fluoro-3-iodo-4-methoxybenzene ≥97%
2-Ethoxy-1-fluoro-3-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO₂
MW: 296.0773 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759986
CAS Number: 2385205-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR