3-Ethoxy-2-fluoro-6-iodobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

60832-500MG 60832-1G 60832-25G 60832-5G
77331-048EA 305.91 USD
77331-048 77355-274 77388-772 77379-348
3-Ethoxy-2-fluoro-6-iodobenzaldehyde ≥95%
3-Ethoxy-2-fluoro-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈FIO₂
MW: 294.0614 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759969
CAS Number: 2385553-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR