(3-Ethoxy-2-fluoro-6-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

60826-500MG 60826-1G 60826-5G
77339-634EA 604.61 USD
77339-634 77355-270 77379-346
(3-Ethoxy-2-fluoro-6-(methylthio)phenyl)methanol ≥97%
(3-Ethoxy-2-fluoro-6-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂S
MW: 216.2724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759963
CAS Number: 2901109-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR