3-Ethoxy-2-fluoro-6-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60825-500MG 60825-1G 60825-5G
77338-614EA 503.67 USD
77338-614 77355-268 77379-344
3-Ethoxy-2-fluoro-6-(methylthio)benzaldehyde ≥97%
3-Ethoxy-2-fluoro-6-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂S
MW: 214.2565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759962
CAS Number: 2807450-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR