(6-Methyl-[1,1'-biphenyl]-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

60811-500MG 60811-1G 60811-5G
77339-408EA 586.07 USD
77339-408 77355-262 77379-338
(6-Methyl-[1,1'-biphenyl]-3-yl)methanol ≥97%
(6-Methyl-[1,1'-biphenyl]-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
MW: 198.2603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19315086
CAS Number: 89951-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR