Ethyl 3-ethyl-4-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60809-500MG 60809-1G 60809-5G
77336-222EA 383.16 USD
77336-222 77355-258 77379-334
Ethyl 3-ethyl-4-formylbenzoate ≥97%
Ethyl 3-ethyl-4-formylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.2377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759954
CAS Number: 2764731-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR