Methyl 4-chloro-2-methyl-2H-indazole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

60806-500MG 60806-1G 60806-5G
77335-464EA 354.32 USD
77335-464 77355-256 77379-332
Methyl 4-chloro-2-methyl-2H-indazole-5-carboxylate ≥97%
Methyl 4-chloro-2-methyl-2H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34759953
CAS Number: 2120737-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR