4-Chloro-2-methyl-2H-indazole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

60805-500MG 60805-1G 60805-5G 60805-25G
77331-044EA 252.35 USD
77331-044 77355-254 77367-234 77388-768
4-Chloro-2-methyl-2H-indazole-5-carboxylic acid ≥97%
4-Chloro-2-methyl-2H-indazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂
MW: 210.6170 g/mol
Storage Temperature: Ambient
CAS Number: 1890523-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR