Methyl 2,6-dichloro-3-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60802-500MG 60802-1G 60802-5G
77337-926EA 451.14 USD
77337-926 77355-250 77379-330
Methyl 2,6-dichloro-3-formylbenzoate ≥97%
Methyl 2,6-dichloro-3-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₃
MW: 233.0481 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34475501
CAS Number: 311348-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR