Ethyl 2,6-dichloro-3-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60801-500MG 60801-1G 60801-5G
77337-924EA 451.14 USD
77337-924 77355-248 77379-328
Ethyl 2,6-dichloro-3-formylbenzoate ≥97%
Ethyl 2,6-dichloro-3-formylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₃
MW: 247.0747 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759949
CAS Number: 311348-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR