Ethyl 2-formyl-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60794-500MG 60794-1G 60794-5G
77331-036EA 319.3 USD
77331-036 77355-246 77379-326
Ethyl 2-formyl-4-methoxybenzoate ≥97%
Ethyl 2-formyl-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.2105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25969869
CAS Number: 1401160-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR