Ethyl 4-formyl-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60791-500MG 60791-1G 60791-5G
77337-776EA 441.87 USD
77337-776 77355-244 77379-324
Ethyl 4-formyl-2,5-dimethylbenzoate ≥97%
Ethyl 4-formyl-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.2377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759942
CAS Number: 2387035-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR