1-Iodo-4-(methoxymethoxy)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

60789-500MG 60789-1G 60789-5G 60789-25G
77331-034EA 175.1 USD
77331-034 77346-536 77367-226 77388-760
1-Iodo-4-(methoxymethoxy)-2-methylbenzene ≥97%
1-Iodo-4-(methoxymethoxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₂
MW: 278.0869 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759941
CAS Number: 1309378-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR