3-Bromo-5-chloro-N-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

60786-500MG 60786-1G 60786-5G 60786-25G
77331-032EA 98.88 USD
77331-032 77342-722 77367-224 77379-322
3-Bromo-5-chloro-N-methoxybenzamide ≥97%
3-Bromo-5-chloro-N-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.5036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30297841
CAS Number: 1329406-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR