Ethyl 4-formyl-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60784-500MG 60784-1G 60784-25G 60784-5G
77331-030EA 293.55 USD
77331-030 77355-242 77388-758 77379-320
Ethyl 4-formyl-3-methylbenzoate ≥97%
Ethyl 4-formyl-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30487182
CAS Number: 203856-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR