2,4,6-Tribromo-3-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

60783-500MG 60783-1G 60783-5G 60783-25G
77331-028EA 251.32 USD
77331-028 77355-240 77367-222 77388-756
2,4,6-Tribromo-3-isopropoxypyridine ≥97%
2,4,6-Tribromo-3-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₈Br₃NO
Storage Temperature: Ambient
MDL Number: MFCD34759940
CAS Number: 2764731-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR