3-(Benzyloxy)-2,4,6-tribromopyridine ≥97%

Supplier: AOBChem USA

Warning

60782-500MG 60782-1G 60782-5G 60782-25G
77331-026EA 226.6 USD
77331-026 77355-238 77367-220 77388-754
3-(Benzyloxy)-2,4,6-tribromopyridine ≥97%
3-(Benzyloxy)-2,4,6-tribromopyridine

Precaución: for Research Use Only

Formula: C₁₂H₈Br₃NO
MW: 421.9100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759939
CAS Number: 2764731-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR