1,5-Dibromo-2-chloro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60756-500MG 60756-1G 60756-5G
77331-018EA 325.48 USD
77331-018 77355-234 77379-314
1,5-Dibromo-2-chloro-4-(trifluoromethyl)benzene ≥97%
1,5-Dibromo-2-chloro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂Br₂ClF₃
MW: 338.3469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759921
CAS Number: 2385812-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR