1,2,4-Tribromo-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

60755-500MG 60755-1G 60755-5G
77331-016EA 316.21 USD
77331-016 77355-232 77379-312
1,2,4-Tribromo-5-(trifluoromethyl)benzene ≥97%
1,2,4-Tribromo-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂Br₃F₃
Storage Temperature: Ambient
MDL Number: MFCD34759920
CAS Number: 2384961-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR