1-(Difluoromethoxy)-4,5-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

60751-500MG 60751-1G 60751-5G
77339-430EA 589.16 USD
77339-430 77355-230 77379-310
1-(Difluoromethoxy)-4,5-difluoro-2-iodobenzene ≥97%
1-(Difluoromethoxy)-4,5-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₇H₃F₄IO
MW: 305.9962 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28788109
CAS Number: 1803833-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR