1-(Benzyloxy)-4,5-difluoro-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

60750-500MG 60750-1G 60750-5G
77331-014EA 347.11 USD
77331-014 77355-228 77379-308
1-(Benzyloxy)-4,5-difluoro-2-iodobenzene ≥95%
1-(Benzyloxy)-4,5-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34648293
CAS Number: 2098626-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR