1,5-Dibromo-2-iodo-4-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

60739-500MG 60739-1G 60739-5G 60739-25G
77331-008EA 139.05 USD
77331-008 77344-672 77367-210 77388-746
1,5-Dibromo-2-iodo-4-(trifluoromethoxy)benzene ≥95%
1,5-Dibromo-2-iodo-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₂Br₂F₃IO
Storage Temperature: Ambient
MDL Number: MFCD34759908
CAS Number: 2386001-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR