5-Bromo-1-propyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

60736-500MG 60736-1G 60736-5G 60736-25G
77331-006EA 175.1 USD
77331-006 77346-494 77367-206 77388-744
5-Bromo-1-propyl-1H-pyrazole ≥97%
5-Bromo-1-propyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD23134838
CAS Number: 1427021-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR