3-Bromo-2,6-dichloroisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

60716-500MG 60716-1G 60716-5G
77339-422EA 588.13 USD
77339-422 77355-220 77379-300
3-Bromo-2,6-dichloroisonicotinic acid ≥95%
3-Bromo-2,6-dichloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34759899
CAS Number: 2807448-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR