2-(2-Methyl-4-(trifluoromethyl)phenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

60711-500MG 60711-1G 60711-5G 60711-25G
77331-004EA 184.37 USD
77331-004 77347-242 77367-204 77388-742
2-(2-Methyl-4-(trifluoromethyl)phenyl)propan-2-ol ≥97%
2-(2-Methyl-4-(trifluoromethyl)phenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃O
MW: 218.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759895
CAS Number: 1599720-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR