4-Chloro-2-fluoro-N,N-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

60698-500MG 60698-1G 60698-5G 60698-25G
77330-998EA 100.94 USD
77330-998 77342-954 77367-198 77379-292
4-Chloro-2-fluoro-N,N-dimethylaniline ≥97%
4-Chloro-2-fluoro-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₉ClFN
MW: 173.615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116327
CAS Number: 914635-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR